missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ IsoClean™ Cleanroom Front Snap Frocks, Bound Neck
Tyvek™ IsoClean™ provides an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash. Clean processed versions available.
Supplier: DuPont™ IC270BWH2X003000
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
- Frocks provide body-length jacket design with front snap closures for easy donning and doffing
- Made of Tyvek™, a spunbonded olefin fabric that is lightweight and flexible
- Processed in a certified ISO Class 4 (formerly Class 10) cleanroom, antistatic treated and packaged in durable, antistatic bags
- Frocks are ISO Class 5 (formerly Class 100) compatible
- Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
- Elastic opening for tighter fit at wrist
- Bound neck for lower particle shedding
- Adjustable snap at neck for better fit
- Clean processed garments are individually packaged
- Garments not clean processed are bulk packaged in double transparent poly liners
Specifications
Controlled Environments Frock | |
Tyvek | |
Single-use | |
Snaps | |
Bound | |
Yes | |
101 to 109 cm | |
Bulk Packaged | |
111.76 cm | |
98.4 cm | |
Class 100 (ISO 5) |
2X-Large | |
White | |
Non-sterile | |
Elastic | |
Bound | |
40 to 43 in. | |
Set-in Sleeve | |
44 in. | |
38.75 in. | |
Not Clean Processed |