missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DuPont™ IsoClean™ Cleanroom Front Snap Frocks, Bound Neck

Tyvek™ IsoClean™ provides an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash. Clean processed versions available.

$315.29 - $588.49

Specifications

Product Type Controlled Environments Frock
Material Tyvek
Color White
Disposable Single-use
Sterility Non-sterile
View More Specs

Alert:

Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.


Products 14
Catalog Number Mfr. No. Size Packaging Type Description Price Quantity  
Catalog Number Mfr. No. Size Packaging Type Description Price Quantity  
29-636-1266
View Documents
DuPont™
IC270BWHSM00300C
Small Individually Packaged Clean Processed
Case of 30 Each for $484.94
 
29-636-1267
View Documents
DuPont™
IC270BWHMD00300C
Medium Individually Packaged Clean Processed
Case of 30 Each for $484.94
 
29-636-1268
View Documents
DuPont™
IC270BWHLG00300C
Large Individually Packaged Clean Processed
Case of 30 Each for $484.94
 
29-636-1269
View Documents
DuPont™
IC270BWHXL00300C
X-Large Individually Packaged Clean Processed
Case of 30 Each for $484.94
 
29-636-1270
View Documents
DuPont™
IC270BWH2X00300C
2X-Large Individually Packaged Clean Processed
Case of 3 Each for $514.04
 
190881125F
View Documents
DuPont™
IC270BWH3X00300C
3X-Large Individually Packaged Clean Processed
Case of 30 Each for $544.86
 
19-088-1124A
View Documents
DuPont™
IC270BWHSM003000
Small Bulk Packaged Not Clean Processed
Case of 30 Each for $315.29
 
19-088-1124B
View Documents
DuPont™
IC270BWHMD003000
Medium Bulk Packaged Not Clean Processed
Case of 30 Each for $315.29
 
19-088-1124C
View Documents
DuPont™
IC270BWHLG003000
Large Bulk Packaged Not Clean Processed
Case of 30 Each for $333.57
 
19-088-1124D
View Documents
DuPont™
IC270BWHXL003000
X-Large Bulk Packaged Not Clean Processed
Case of 30 Each for $315.29
 
19-088-1124E
View Documents
DuPont™
IC270BWH2X003000
2X-Large Bulk Packaged Not Clean Processed
Case of 30 Each for $334.16
 
19-088-1124F
View Documents
DuPont™
IC270BWH3X003000
3X-Large Bulk Packaged Not Clean Processed
Case of 30 Each for $354.20
 
17-988-085
View Documents
DuPont™
IC270BWH4X003000
4X-Large Bulk Packaged Not Clean Processed
Case of 30 Each for $382.49
 
19-088-1125G
View Documents
DuPont™
IC270BWH4X00300C
4X-Large Individually Packaged Clean Processed
Case of 30 Each for $588.49
 
Description

Description

  • Frocks provide body-length jacket design with front snap closures for easy donning and doffing
  • Made of Tyvek™, a spunbonded olefin fabric that is lightweight and flexible
  • Processed in a certified ISO Class 4 (formerly Class 10) cleanroom, antistatic treated and packaged in durable, antistatic bags
  • Frocks are ISO Class 5 (formerly Class 100) compatible
  • Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
  • Elastic opening for tighter fit at wrist
  • Bound neck for lower particle shedding
  • Adjustable snap at neck for better fit
  • Clean processed garments are individually packaged
  • Garments not clean processed are bulk packaged in double transparent poly liners
Specifications

Specifications

Controlled Environments Frock
White
Non-sterile
Elastic
Bound
Set-in Sleeve
Tyvek
Single-use
Snaps
Bound
Yes
Class 100 (ISO 5)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'