missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isonicotinic acid, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 122591000

 View more versions of this product

Catalog No. AC122591000

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Isonicotinic acid
310°C to 315°C
260°C (15.0 mmHg)
Plastic bottle
100 g
15,5233
Solubility in water: 5.2g/L (20°C). Other solubilities: practically insoluble in benzene,ether and,boiling alcohol
C1=CN=CC=C1C(=O)O
123.11
CHEBI:6032
99%
55-22-1
Cream
99%
C6H5NO2
22,45
isonicotinic acid, 4-pyridinecarboxylic acid, 4-picolinic acid, 4-carboxypyridine, p-pyridinecarboxylic acid, gamma-picolinic acid, gamma-pyridinecarboxylic acid, acide iso-nicotinique, 1,4-dihydroisonicotinic acid, acide iso-nicotinique french
TWBYWOBDOCUKOW-UHFFFAOYSA-N
pyridine-4-carboxylic acid
5922
123.11
Powder

Chemical Identifiers

55-22-1
123.11
isonicotinic acid, 4-pyridinecarboxylic acid, 4-picolinic acid, 4-carboxypyridine, p-pyridinecarboxylic acid, gamma-picolinic acid, gamma-pyridinecarboxylic acid, acide iso-nicotinique, 1,4-dihydroisonicotinic acid, acide iso-nicotinique french
CHEBI:6032
C1=CN=CC=C1C(=O)O
C6H5NO2
TWBYWOBDOCUKOW-UHFFFAOYSA-N
5922
pyridine-4-carboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:

GHS Signal Word: Warning

EINECSNumber : 200-228-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only