missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isonicotinic acid, 99%, Thermo Scientific Chemicals

$48.36 - $532.05

Chemical Identifiers

CAS 55-22-1
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.11
InChI Key TWBYWOBDOCUKOW-UHFFFAOYSA-N
Synonym isonicotinic acid, 4-pyridinecarboxylic acid, 4-picolinic acid, 4-carboxypyridine, p-pyridinecarboxylic acid, gamma-picolinic acid, gamma-pyridinecarboxylic acid, acide iso-nicotinique, 1,4-dihydroisonicotinic acid, acide iso-nicotinique french
PubChem CID 5922
ChEBI CHEBI:6032
IUPAC Name pyridine-4-carboxylic acid
SMILES C1=CN=CC=C1C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC122591000
SDS View Documents
Thermo Scientific Chemicals
122591000
100 g Plastic bottle
Each for $48.36
 
AC122595000
SDS View Documents
Thermo Scientific Chemicals
122595000
500 g Plastic bottle
Each for $137.92
 
AC122590025
SDS View Documents
Thermo Scientific Chemicals
122590025
2.5 kg Plastic bottle
Each for $532.05
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

55-22-1
123.11
isonicotinic acid, 4-pyridinecarboxylic acid, 4-picolinic acid, 4-carboxypyridine, p-pyridinecarboxylic acid, gamma-picolinic acid, gamma-pyridinecarboxylic acid, acide iso-nicotinique, 1,4-dihydroisonicotinic acid, acide iso-nicotinique french
CHEBI:6032
C1=CN=CC=C1C(=O)O
C6H5NO2
TWBYWOBDOCUKOW-UHFFFAOYSA-N
5922
pyridine-4-carboxylic acid

Specifications

55-22-1
Cream
99%
C6H5NO2
22,45
isonicotinic acid, 4-pyridinecarboxylic acid, 4-picolinic acid, 4-carboxypyridine, p-pyridinecarboxylic acid, gamma-picolinic acid, gamma-pyridinecarboxylic acid, acide iso-nicotinique, 1,4-dihydroisonicotinic acid, acide iso-nicotinique french
TWBYWOBDOCUKOW-UHFFFAOYSA-N
pyridine-4-carboxylic acid
5922
123.11
Powder
310°C to 315°C
260°C (15.0 mmHg)
Plastic bottle
100 g
15,5233
Solubility in water: 5.2g/L (20°C). Other solubilities: practically insoluble in benzene,ether and,boiling alcohol
C1=CN=CC=C1C(=O)O
123.11
CHEBI:6032
99%
Isonicotinic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:

GHS Signal Word: Warning

EINECSNumber : 200-228-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only