missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isoprene, 98%, stabilized, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 122671000

 View more versions of this product

Catalog No. AC122671000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Isoprene
-146.0°C
0.6800g/mL
−48°C
97.5% min. (GC)
C5H8
H2C=CHC(CH3)=CH2
100 mL
13,196
15, 5247
Solubility in water: 0.7g/L (25°C). Other solubilities: miscible with alcohol,ether and acetone,oils/fat
CC(=C)C=C
68.11
6557
CHEBI:35194
98%
78-79-5,98-29-3
Colorless to Yellow
34.0°C
Authentic
Glass bottle
1.4205 to 1.4225
MFCD00008600
01, 252
0.68
isoprene, 2-methyl-1,3-butadiene, isopentadiene, 2-methylbutadiene, 1,3-butadiene, 2-methyl, 2-methyldivinyl, beta-methylbivinyl, isopreno, isoterpene, isopren
RRHGJUQNOFWUDK-UHFFFAOYSA-N
2-methylbuta-1,3-diene
100 to 250ppm 4-tert-butylcatechol
0.225 mPa.s (15°C)
68.11
Liquid

Chemical Identifiers

78-79-5
68.11
RRHGJUQNOFWUDK-UHFFFAOYSA-N
6557
2-methylbuta-1,3-diene
C5H8
MFCD00008600
isoprene, 2-methyl-1,3-butadiene, isopentadiene, 2-methylbutadiene, 1,3-butadiene, 2-methyl, 2-methyldivinyl, beta-methylbivinyl, isopreno, isoterpene, isopren
CHEBI:35194
CC(=C)C=C
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause cancer.
Suspected of causing genetic defects.
Toxic to aquatic life with long lasting effects.
Extremely flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Obtain special instructions before use.
Wea

GHS Signal Word: Danger

EINECSNumber : 201-143-3

RTECSNumber : NT4037000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only