Learn More
Isoprene, 98%, stabilized, Thermo Scientific Chemicals
$66.34 - $173.68
Chemical Identifiers
CAS | 78-79-5 |
---|---|
Molecular Formula | C5H8 |
Molecular Weight (g/mol) | 68.11 |
MDL Number | MFCD00008600 |
InChI Key | RRHGJUQNOFWUDK-UHFFFAOYSA-N |
Synonym | isoprene, 2-methyl-1,3-butadiene, isopentadiene, 2-methylbutadiene, 1,3-butadiene, 2-methyl, 2-methyldivinyl, beta-methylbivinyl, isopreno, isoterpene, isopren |
PubChem CID | 6557 |
ChEBI | CHEBI:35194 |
IUPAC Name | 2-methylbuta-1,3-diene |
SMILES | CC(=C)C=C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC122671000
|
Thermo Scientific Chemicals
122671000 |
100 mL | Glass bottle |
Each for $66.34
|
|
||||
AC122675000
|
Thermo Scientific Chemicals
122675000 |
500 mL | Glass bottle |
Each for $94.75
|
|
||||
AC122670010
|
Thermo Scientific Chemicals
122670010 |
1 L | Glass bottle |
Each for $173.68
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
78-79-5 | |
68.11 | |
RRHGJUQNOFWUDK-UHFFFAOYSA-N | |
6557 | |
2-methylbuta-1,3-diene |
C5H8 | |
MFCD00008600 | |
isoprene, 2-methyl-1,3-butadiene, isopentadiene, 2-methylbutadiene, 1,3-butadiene, 2-methyl, 2-methyldivinyl, beta-methylbivinyl, isopreno, isoterpene, isopren | |
CHEBI:35194 | |
CC(=C)C=C |
Specifications
78-79-5,98-29-3 | |
Colorless to Yellow | |
34.0°C | |
Authentic | |
Glass bottle | |
1.4205 to 1.4225 | |
MFCD00008600 | |
01, 252 | |
0.68 | |
isoprene, 2-methyl-1,3-butadiene, isopentadiene, 2-methylbutadiene, 1,3-butadiene, 2-methyl, 2-methyldivinyl, beta-methylbivinyl, isopreno, isoterpene, isopren | |
RRHGJUQNOFWUDK-UHFFFAOYSA-N | |
2-methylbuta-1,3-diene | |
100 to 250ppm 4-tert-butylcatechol | |
0.225 mPa.s (15°C) | |
68.11 | |
Liquid |
-146.0°C | |
0.6800g/mL | |
−48°C | |
97.5% min. (GC) | |
C5H8 | |
H2C=CHC(CH3)=CH2 | |
100 mL | |
13,196 | |
15, 5247 | |
Solubility in water: 0.7g/L (25°C). Other solubilities: miscible with alcohol,ether and acetone,oils/fat | |
CC(=C)C=C | |
68.11 | |
6557 | |
CHEBI:35194 | |
98% | |
Isoprene |
Safety and Handling
GHS H Statement
May cause cancer.
Suspected of causing genetic defects.
Toxic to aquatic life with long lasting effects.
Extremely flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Obtain special instructions before use.
Wea
GHS Signal Word: Danger
EINECSNumber : 201-143-3
RTECSNumber : NT4037000
TSCA : TSCA
RUO – Research Use Only