Learn More
Isopropyl Alcohol, Certified, 70% (v/v), LabChem™
Supplier: LabChem LC157606
Specifications
Isopropyl Alcohol | |
67-63-0,7732-18-5 | |
70,30 | |
0.88g/mL | |
10 L | |
Passes Test | |
Liquid | |
C3H8O | |
2.5316mm2/s | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
propan-2-ol | |
3776 | |
60.09 | |
Certified |
-88°C | |
Colorless | |
Carbon dioxide; Carbon monoxide | |
82°C | |
12°C | |
Cubitainer | |
0.877g/mL ±0.005 | |
C3H8O | |
UN1219 | |
Soluble in water | |
CC(C)O | |
60.096 | |
CHEBI:17824 | |
70% (v/v) |
Chemical Identifiers
67-63-0 | |
60.096 | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
CHEBI:17824 | |
CC(C)O |
C3H8O | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
3776 | |
propan-2-ol |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.
GHS P Statement
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, lighting, ventilating equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Avoid breathing mist, spray, vapors.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Call a poison center/doctor if you feel unwell.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
In case of fire: Use dry chemical powder, alcohol-resistant foam, carbon dioxide to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
EINECSNumber : 200-661-7
Recommended Storage : Room Temperature