missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Isopropyl Alcohol, Certified, 70% (v/v), LabChem™
$51.85 - $3939.74
Chemical Identifiers
CAS | 67-63-0 |
---|---|
Molecular Formula | C3H8O |
Molecular Weight (g/mol) | 60.096 |
InChI Key | KFZMGEQAYNKOFK-UHFFFAOYSA-N |
Synonym | isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol |
PubChem CID | 3776 |
ChEBI | CHEBI:17824 |
IUPAC Name | propan-2-ol |
SMILES | CC(C)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
LC157601
|
LabChem
LC157601 |
500 mL | Poly Bottle |
Each for $51.85
|
|
||||
LC157602
|
LabChem
LC157602 |
1 L | Poly Bottle |
Each for $68.65
|
|
||||
LC157604
|
LabChem
LC157604 |
4 L | Poly Bottle |
Each for $140.79
|
|
||||
LC157606
|
LabChem
LC157606 |
10 L | Cubitainer |
Each for $340.44
|
|
||||
LC157605
|
LabChem
LC157605 |
20 L | Cubitainer |
Each for $502.70
|
|
||||
LC15760200
|
LabChem
LC15760200 |
200 L | Poly Drum |
Each for $3,939.74
|
|
||||
Chemical Identifiers
67-63-0 | |
60.096 | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
CHEBI:17824 | |
CC(C)O |
C3H8O | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
3776 | |
propan-2-ol |
Specifications
-88°C | |
Colorless | |
Carbon dioxide; Carbon monoxide | |
82°C | |
12°C | |
Poly Bottle | |
C3H8O | |
2.5316mm2/s | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
propan-2-ol | |
3776 | |
60.09 | |
Certified |
67-63-0,7732-18-5 | |
70,30 | |
0.88g/mL | |
500 mL | |
Passes Test | |
Liquid | |
C3H8O | |
UN1219 | |
Soluble in water | |
CC(C)O | |
60.096 | |
CHEBI:17824 | |
70% (v/v) | |
Isopropyl Alcohol |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.
GHS P Statement
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, lighting, ventilating equipment.
Danger
EINECSNumber : 200-661-7
Recommended Storage : Room Temperature