missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Isopropylamine 99.0+%, TCI America™
$27.68 - $38.06
Chemical Identifiers
CAS | 75-31-0 |
---|---|
Molecular Formula | C3H9N |
Molecular Weight (g/mol) | 59.112 |
MDL Number | MFCD00008082 |
InChI Key | JJWLVOIRVHMVIS-UHFFFAOYSA-N |
Synonym | isopropylamine, 2-propanamine, 2-aminopropane, monoisopropylamine, 2-propylamine, sec-propylamine, 1-methylethylamine, isopropyl amine, isopropilamina, propane, 2-amino |
PubChem CID | 6363 |
ChEBI | CHEBI:15739 |
IUPAC Name | propan-2-amine |
SMILES | CC(C)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
I016525ML
|
TCI America
I016525ML |
25 mL |
Each for $27.68
|
|
|||||
I0165500ML
|
TCI America
I0165500ML |
500 mL |
Each for $38.06
|
|
|||||
Chemical Identifiers
75-31-0 | |
59.112 | |
JJWLVOIRVHMVIS-UHFFFAOYSA-N | |
6363 | |
propan-2-amine |
C3H9N | |
MFCD00008082 | |
isopropylamine, 2-propanamine, 2-aminopropane, monoisopropylamine, 2-propylamine, sec-propylamine, 1-methylethylamine, isopropyl amine, isopropilamina, propane, 2-amino | |
CHEBI:15739 | |
CC(C)N |
Specifications
75-31-0 | |
Colorless | |
C3H9N | |
25 mL | |
isopropylamine, 2-propanamine, 2-aminopropane, monoisopropylamine, 2-propylamine, sec-propylamine, 1-methylethylamine, isopropyl amine, isopropilamina, propane, 2-amino | |
CC(C)N | |
59.112 | |
CHEBI:15739 | |
≥99.0% (GC,T) | |
Isopropylamine |
-95°C | |
34°C | |
MFCD00008082 | |
1221 | |
JJWLVOIRVHMVIS-UHFFFAOYSA-N | |
propan-2-amine | |
6363 | |
59.11 | |
Liquid |
Safety and Handling
EINECSNumber : (2)-0131
RTECSNumber : NT8400000
TSCA : Yes