missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isosorbide dimethyl ether, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 369911000

 View more versions of this product

Catalog No. AC369911000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Isosorbide dimethyl ether
Colorless to Yellow
93°C to 95°C (0.1 hPa)
Authentic
Liquid
98.5% min. (GC)
C8H14O4
19, IV, 991
Solubility in water: soluble. Other solubilities: soluble in alcohols
COC1COC2C(COC12)OC
174.20
174.19
5306-85-4
1.1700g/mL
100 g
Glass bottle
1.4600 to 1.4620
1.17
MFCD00064828
dimethyl isosorbide, isosorbide dimethyl ether, 3r,3ar,6s,6ar-3,6-dimethoxyhexahydrofuro 3,2-b furan, unii-sa6a6v432s, 1,4:3,6-dianhydro-2,5-di-o-methyl-d-glucitol, arlasolve dmi, 2,5-di-o-methyl-1,4:3,6-dianhydro-d-glucitol, dimethyl isosorbide ether, 3r,3ar,6s,6ar-3,6-dimethoxy-hexahydrofuro 3,2-b furan, 3s,3ar,6r,6ar-3,6-dimethoxy-2,3,3a,5,6,6a-hexahydrofuro 3,2-b furan
MEJYDZQQVZJMPP-UHFFFAOYNA-N
(3S,3aR,6R,6aR)-3,6-dimethoxy-2,3,3a,5,6,6a-hexahydrofuro[3,2-b]furan
62990
99%

Chemical Identifiers

5306-85-4
174.20
MEJYDZQQVZJMPP-UHFFFAOYNA-N
62990
C8H14O4
MFCD00064828
dimethyl isosorbide, isosorbide dimethyl ether, 3r,3ar,6s,6ar-3,6-dimethoxyhexahydrofuro 3,2-b furan, unii-sa6a6v432s, 1,4:3,6-dianhydro-2,5-di-o-methyl-d-glucitol, arlasolve dmi, 2,5-di-o-methyl-1,4:3,6-dianhydro-d-glucitol, dimethyl isosorbide ether, 3r,3ar,6s,6ar-3,6-dimethoxy-hexahydrofuro 3,2-b furan, 3s,3ar,6r,6ar-3,6-dimethoxy-2,3,3a,5,6,6a-hexahydrofuro 3,2-b furan
COC1COC2C(COC12)OC
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 226-159-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only