missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Isosorbide dimethyl ether, 99%, Thermo Scientific Chemicals
$104.67 - $344.16
Chemical Identifiers
CAS | 5306-85-4 |
---|---|
Molecular Formula | C8H14O4 |
Molecular Weight (g/mol) | 174.20 |
MDL Number | MFCD00064828 |
InChI Key | MEJYDZQQVZJMPP-UHFFFAOYNA-N |
Synonym | dimethyl isosorbide, isosorbide dimethyl ether, 3r,3ar,6s,6ar-3,6-dimethoxyhexahydrofuro 3,2-b furan, unii-sa6a6v432s, 1,4:3,6-dianhydro-2,5-di-o-methyl-d-glucitol, arlasolve dmi, 2,5-di-o-methyl-1,4:3,6-dianhydro-d-glucitol, dimethyl isosorbide ether, 3r,3ar,6s,6ar-3,6-dimethoxy-hexahydrofuro 3,2-b furan, 3s,3ar,6r,6ar-3,6-dimethoxy-2,3,3a,5,6,6a-hexahydrofuro 3,2-b furan |
PubChem CID | 62990 |
SMILES | COC1COC2C(COC12)OC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC369910250
|
Thermo Scientific Chemicals
369910250 |
25 g | Glass bottle |
N/A
|
|
||||
AC369911000
|
Thermo Scientific Chemicals
369911000 |
100 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5306-85-4 | |
174.20 | |
MEJYDZQQVZJMPP-UHFFFAOYNA-N | |
62990 |
C8H14O4 | |
MFCD00064828 | |
dimethyl isosorbide, isosorbide dimethyl ether, 3r,3ar,6s,6ar-3,6-dimethoxyhexahydrofuro 3,2-b furan, unii-sa6a6v432s, 1,4:3,6-dianhydro-2,5-di-o-methyl-d-glucitol, arlasolve dmi, 2,5-di-o-methyl-1,4:3,6-dianhydro-d-glucitol, dimethyl isosorbide ether, 3r,3ar,6s,6ar-3,6-dimethoxy-hexahydrofuro 3,2-b furan, 3s,3ar,6r,6ar-3,6-dimethoxy-2,3,3a,5,6,6a-hexahydrofuro 3,2-b furan | |
COC1COC2C(COC12)OC |
Specifications
5306-85-4 | |
1.1700g/mL | |
25 g | |
Glass bottle | |
1.4600 to 1.4620 | |
1.17 | |
MFCD00064828 | |
dimethyl isosorbide, isosorbide dimethyl ether, 3r,3ar,6s,6ar-3,6-dimethoxyhexahydrofuro 3,2-b furan, unii-sa6a6v432s, 1,4:3,6-dianhydro-2,5-di-o-methyl-d-glucitol, arlasolve dmi, 2,5-di-o-methyl-1,4:3,6-dianhydro-d-glucitol, dimethyl isosorbide ether, 3r,3ar,6s,6ar-3,6-dimethoxy-hexahydrofuro 3,2-b furan, 3s,3ar,6r,6ar-3,6-dimethoxy-2,3,3a,5,6,6a-hexahydrofuro 3,2-b furan | |
MEJYDZQQVZJMPP-UHFFFAOYNA-N | |
3,6-dimethoxy-hexahydrofuro[3,2-b]furan | |
62990 | |
99% |
Colorless to Yellow | |
93°C to 95°C (0.1 hPa) | |
Authentic | |
Liquid | |
98.5% min. (GC) | |
C8H14O4 | |
19, IV, 991 | |
Solubility in water: soluble. Other solubilities: soluble in alcohols | |
COC1COC2C(COC12)OC | |
174.20 | |
174.19 | |
Isosorbide dimethyl ether |
Safety and Handling
EINECSNumber : 226-159-8
RUO – Research Use Only