Learn More
Isovaleric acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 156691000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Isovaleric acid | |
503-74-2 | |
Colorless to Yellow | |
176.0°C | |
Authentic | |
Glass bottle | |
1.3990 to 1.4070 | |
MFCD00002726 | |
02, 309 | |
15, 5277 | |
Solubility in water: 25g/L (20°C). Other solubilities: soluble in alcohol,chloroform and ether | |
CC(C)CC(O)=O | |
102.13 | |
2.4 mPa.s (20°C) | |
102.13 | |
Liquid |
99% | |
-35.0°C | |
0.9260g/mL | |
70°C | |
98.5% min. (GC) | |
C5H10O2 | |
(CH3)2CHCH2CO2H | |
100 g | |
0.926 | |
isovaleric acid, isopentanoic acid, 3-methylbutyric acid, delphinic acid, isopropylacetic acid, butanoic acid, 3-methyl, isovalerianic acid, isobutylformic acid, 3-methylbutyrate, beta-methylbutyric acid | |
GWYFCOCPABKNJV-UHFFFAOYSA-N | |
3-methylbutanoic acid | |
10430 | |
CHEBI:28484 | |
99% |
Chemical Identifiers
503-74-2 | |
102.13 | |
GWYFCOCPABKNJV-UHFFFAOYSA-N | |
10430 | |
3-methylbutanoic acid |
C5H10O2 | |
MFCD00002726 | |
isovaleric acid, isopentanoic acid, 3-methylbutyric acid, delphinic acid, isopropylacetic acid, butanoic acid, 3-methyl, isovalerianic acid, isobutylformic acid, 3-methylbutyrate, beta-methylbutyric acid | |
CHEBI:28484 | |
CC(C)CC(O)=O |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 207-975-3
RTECSNumber : NY1400000
TSCA : TSCA
RUO – Research Use Only