missing translation for 'onlineSavingsMsg'
Learn More

Isovaleric acid, 99%

CAS: 503-74-2 | C5H10O2 | 102.13 g/mol

$74.29 - $136.21

Chemical Identifiers

CAS 503-74-2
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00002726
InChI Key GWYFCOCPABKNJV-UHFFFAOYSA-N
Synonym isovaleric acid, isopentanoic acid, 3-methylbutyric acid, delphinic acid, isopropylacetic acid, butanoic acid, 3-methyl, isovalerianic acid, isobutylformic acid, 3-methylbutyrate, beta-methylbutyric acid
PubChem CID 10430
ChEBI CHEBI:28484
IUPAC Name 3-methylbutanoic acid
SMILES CC(C)CC(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC156690100
SDS View Documents
Thermo Scientific Chemicals
156690100
10 g Glass bottle
Each for $74.29
Only null left
 
AC156691000
SDS View Documents
Thermo Scientific Chemicals
156691000
100 g Glass bottle
Each for $81.16
Only null left
 
AC156695000
SDS View Documents
Thermo Scientific Chemicals
156695000
500 g Glass bottle
Each for $136.21
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

503-74-2
102.13
GWYFCOCPABKNJV-UHFFFAOYSA-N
10430
3-methylbutanoic acid
C5H10O2
MFCD00002726
isovaleric acid, isopentanoic acid, 3-methylbutyric acid, delphinic acid, isopropylacetic acid, butanoic acid, 3-methyl, isovalerianic acid, isobutylformic acid, 3-methylbutyrate, beta-methylbutyric acid
CHEBI:28484
CC(C)CC(O)=O

Specifications

503-74-2
Colorless to Yellow
176.0°C
Authentic
Glass bottle
1.3990 to 1.4070
MFCD00002726
02, 309
15, 5277
Solubility in water: 25g/L (20°C). Other solubilities: soluble in alcohol,chloroform and ether
CC(C)CC(O)=O
102.13
2.4 mPa.s (20°C)
102.13
Liquid
-35.0°C
0.9260g/mL
70°C
98.5% min. (GC)
C5H10O2
(CH3)2CHCH2CO2H
10 g
0.926
isovaleric acid, isopentanoic acid, 3-methylbutyric acid, delphinic acid, isopropylacetic acid, butanoic acid, 3-methyl, isovalerianic acid, isobutylformic acid, 3-methylbutyrate, beta-methylbutyric acid
GWYFCOCPABKNJV-UHFFFAOYSA-N
3-methylbutanoic acid
10430
CHEBI:28484
99%
Isovaleric acid, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 207-975-3

RTECSNumber : NY1400000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only