missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isovaleric acid, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 156690100

 View more versions of this product

Catalog No. AC156690100


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Isovaleric acid
503-74-2
Colorless to Yellow
176.0°C
Authentic
Glass bottle
1.3990 to 1.4070
MFCD00002726
02, 309
15, 5277
Solubility in water: 25g/L (20°C). Other solubilities: soluble in alcohol,chloroform and ether
CC(C)CC(O)=O
102.13
2.4 mPa.s (20°C)
102.13
Liquid
99%
-35.0°C
0.9260g/mL
70°C
98.5% min. (GC)
C5H10O2
(CH3)2CHCH2CO2H
10 g
0.926
isovaleric acid, isopentanoic acid, 3-methylbutyric acid, delphinic acid, isopropylacetic acid, butanoic acid, 3-methyl, isovalerianic acid, isobutylformic acid, 3-methylbutyrate, beta-methylbutyric acid
GWYFCOCPABKNJV-UHFFFAOYSA-N
3-methylbutanoic acid
10430
CHEBI:28484
99%

Chemical Identifiers

503-74-2
102.13
GWYFCOCPABKNJV-UHFFFAOYSA-N
10430
3-methylbutanoic acid
C5H10O2
MFCD00002726
isovaleric acid, isopentanoic acid, 3-methylbutyric acid, delphinic acid, isopropylacetic acid, butanoic acid, 3-methyl, isovalerianic acid, isobutylformic acid, 3-methylbutyrate, beta-methylbutyric acid
CHEBI:28484
CC(C)CC(O)=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 207-975-3

RTECSNumber : NY1400000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only