Learn More
Itraconazole, 98%, Thermo Scientific Chemicals
$163.10 - $1413.10
Chemical Identifiers
CAS | 84625-61-6 |
---|---|
Molecular Formula | C35H38Cl2N8O4 |
Molecular Weight (g/mol) | 705.64 |
MDL Number | MFCD00870168,MFCD00941396 |
InChI Key | VHVPQPYKVGDNFY-ZPGVKDDISA-N |
Synonym | Oriconazole; R51211 |
PubChem CID | 45039617 |
IUPAC Name | 1-(butan-2-yl)-4-{4-[4-(4-{[(2R,4S)-2-(2,4-dichlorophenyl)-2-[(1H-1,2,4-triazol-1-yl)methyl]-1,3-dioxolan-4-yl]methoxy}phenyl)piperazin-1-yl]phenyl}-4,5-dihydro-1H-1,2,4-triazol-5-one |
SMILES | CCC(C)N1N=CN(C1=O)C1=CC=C(C=C1)N1CCN(CC1)C1=CC=C(OC[C@H]2CO[C@@](CN3C=NC=N3)(O2)C2=CC=C(Cl)C=C2Cl)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6639003
|
Thermo Scientific Chemicals
J6639003 |
1 g |
Each for $163.10
|
|
|||||
AAJ6639006
|
Thermo Scientific Chemicals
J6639006 |
5 g |
Each for $319.95
|
|
|||||
AAJ6639014
|
Thermo Scientific Chemicals
J6639014 |
25 g |
Each for $1,413.10
|
|
|||||
Description
Itraconazole is a triazole antifungal agent. It is used to inhibit cytochrome P-450-dependent enzymes and ergosterol synthesis. It has been used against histoplasmosis, blastomycosis, cryptococcal meningitis, and aspergillosis. It?s different formulations are used to study Candida strains in murine invasive infections. It has been used to study proliferative changes of the forestomach mucosa in alloxan-induced diabetic rats..
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsItraconazole is a triazole antifungal agent. It is used to inhibit cytochrome P-450-dependent enzymes and ergosterol synthesis. It has been used against histoplasmosis, blastomycosis, cryptococcal meningitis, and aspergillosis. It′s different formulations are used to study Candida strains in murine invasive infections. It has been used to study proliferative changes of the forestomach mucosa in alloxan-induced diabetic rats..
Solubility
Insoluble in water. Solube in chloroform at 50 mg/ml. Slightly soluble in ethanol or methanol
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Chemical Identifiers
84625-61-6 | |
705.64 | |
VHVPQPYKVGDNFY-ZPGVKDDISA-N | |
45039617 | |
CCC(C)N1N=CN(C1=O)C1=CC=C(C=C1)N1CCN(CC1)C1=CC=C(OC[C@H]2CO[C@@](CN3C=NC=N3)(O2)C2=CC=C(Cl)C=C2Cl)C=C1 |
C35H38Cl2N8O4 | |
MFCD00870168,MFCD00941396 | |
Oriconazole; R51211 | |
1-(butan-2-yl)-4-{4-[4-(4-{[(2R,4S)-2-(2,4-dichlorophenyl)-2-[(1H-1,2,4-triazol-1-yl)methyl]-1,3-dioxolan-4-yl]methoxy}phenyl)piperazin-1-yl]phenyl}-4,5-dihydro-1H-1,2,4-triazol-5-one |
Specifications
Itraconazole | |
84625-61-6 | |
White | |
>110°C (230°F) | |
MFCD00870168,MFCD00941396 | |
Oriconazole; R51211 | |
VHVPQPYKVGDNFY-ZPGVKDDISA-N | |
1-(butan-2-yl)-4-{4-[4-(4-{[(2R,4S)-2-(2,4-dichlorophenyl)-2-[(1H-1,2,4-triazol-1-yl)methyl]-1,3-dioxolan-4-yl]methoxy}phenyl)piperazin-1-yl]phenyl}-4,5-dihydro-1H-1,2,4-triazol-5-one | |
45039617 | |
98% |
Powder | |
166°C | |
1 g | |
C35H38Cl2N8O4 | |
14,5245 | |
Insoluble in water; Solube in chloroform at 50mg/ml; Slightly soluble in ethanol or methanol | |
CCC(C)N1N=CN(C1=O)C1=CC=C(C=C1)N1CCN(CC1)C1=CC=C(OC[C@H]2CO[C@@](CN3C=NC=N3)(O2)C2=CC=C(Cl)C=C2Cl)C=C1 | |
705.64 | |
705.63 | |
Itraconazole |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
RTECSNumber : XZ5481000
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only