JAC Medical Mobile Phlebotomy Carts
Compact ergonomic design pulls up to patient bedside
Supplier: JAC Medical JAC151
Description
- Durable stainless-steel frame
- Five equilateral legs for stability and maintenance-free casters for mobility
- Knob on cart adjusts height from 30 to 49 in. (76.2 to 124.5cm)
- Autoclavable trays and components can be removed from top of cart and carried by hand
- Available with midsize or large trays, either empty or with full tray components; accessories included with select carts
- Optional accessories available for cart customization
Specifications
Phlebotomy Cart | |
76.2 to 124.5 cm | |
Stainless Steel |
30 to 49 in. | |
Large tray for 65+ tubes (with tray components) |