JAC Medical Mobile Phlebotomy Carts
Compact ergonomic design pulls up to patient bedside
Supplier:  JAC Medical JAC155WSH
 View more versions of this product
Catalog No. 22-038-4546
Explore available promotions
Description
Description
  • Durable stainless-steel frame
  • Five equilateral legs for stability and maintenance-free casters for mobility
  • Knob on cart adjusts height from 30 to 49 in. (76.2 to 124.5cm)
  • Autoclavable trays and components can be removed from top of cart and carried by hand
  • Available with midsize or large trays, either empty or with full tray components; accessories included with select carts
  • Optional accessories available for cart customization
Specifications
Specifications
RTD Probe, 12 L x 0.18 in. dia., Stainless Steel, 4-wires Configuration, 100ohm(s) Precision, With 3
76.2 to 124.5 cm
Stainless Steel
30 to 49 in.
Large tray for 65+ tubes (with tray components), writing surface, large equipment basket and bracket, 1 qt. sharps bracket, 3.25 gal. wastebasket and bracket