Justrite™ Cylinder Storage Locker
A Justrite Cylinder Storage Locker provides secure vertical storage for high-pressure compressed gas cylinders and LPG containers.
Supplier: Justrite™ JMC23006
Description
- Heavy-gauge aluminum mesh and rigid extruded frame won't rust, discolor, or corrode
- Sloped roof design sheds water and ice
- Mesh sides prevent buildup of flammable gases and allow easy visual inspection of contents
- Cylinders are kept clear of the ground with strong floor plate
Order Info
Come padlock-ready for security. Predrilled 0.5 in. (1.3cm) anchor holes for stability. Shipped fully assembled.
Compliance
Meet NFPA 58 and OSHA 1910.110 requirements.
Specifications
Exclusive Solid Aluminum roof panel, 2 warning and identification labels | |
Storage Cabinet | |
165.1 cm | |
65 in. | |
Aluminum | |
76 cm |
32 in. | |
81.3 cm | |
Manual-close | |
30 in. | |
NFPA, OSHA |