missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Justrite™ Sure-Grip™ EX Flammable Safety Cabinet

When using a Sure-Grip EX Flammable Safety Cabinets, these two-door and bifold door cabinets protect workers, reduce fire risks, and improve productivity by allowing storage of flammable fuels and chemicals in code-compliant, sturdy 18-gauge steel structures.

Supplier:  Justrite™ 894520

 View more versions of this product

Catalog No. 19-120-2336


Explore available promotions

Description

Description

Features of Justrite Flammables Cabinets

  • Double-wall construction provides 1.5 in. of insulating air space to help prevent the spread of flames
  • High-visibility Haz-Alert™ reflective warning labels offer added safety during power outages or fires
  • Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away
  • Easy-closing, self-latching doors facilitate fire protection of the cabinet
  • Self latch engages critical three-point latching system for protection under fire conditions
  • Fusible links hold self-closing doors wide open, flush with front of cabinet for easy access and resupply; automatically release and slam doors shut at 74°C (165°F)
  • Concealed self-close mechanism provides optimum storage space and obstruction-free access to contents

  • SpillSlope™ safety shelves direct incidental spills to the back and bottom of leakproof sump
  • Adjustable leveling feet ensure solid positioning and alignment for tight door closure
  • Exterior side panel has built-in grounding connector for easy grounding
  • Finished inside and out with durable lead-free powder paint
  • Lettering on door reads: “FLAMMABLE—KEEP FIRE AWAY”
  • Accepts padlock (not included)
  • Color: yellow, gray (optional), white (19-160-376)

Certifications

FM approved.

Compliance

Meet NFPA Code 30 and comply with OSHA regulations.

Warranty

10 years

Specifications

Specifications

100.3 cm
65 in.
Steel (CR)
178 kg
14.56 in.
45.7 cm
45 gal.
39.5 in.
153.7 cm
Flammable liquid storage and protection
109 cm
165.1 cm
2
37 cm
Epoxy, Polyester
U-Loc™ padlockable handle, Haz-Alert™ reflective trilingual warning labels, sturdy SpillSlope™ shelves, three-point Stainless Steel bullet self latching system and welded construction, dual vents with
18 in.
14.56 x 39.5 x 60.5 in. (37 x 100.3 x 153.7 cm)
392 lb.
2
60.5 in.
Can storage cabinet
43 in.
Yellow
Self-close
FM, NFPA, OSHA
Videos
SDS
Documents

Documents

Promotions

Promotions