missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Justrite™ Sure-Grip™ EX Flammable Safety Cabinet
When using a Sure-Grip EX Flammable Safety Cabinets, these two-door and bifold door cabinets protect workers, reduce fire risks, and improve productivity by allowing storage of flammable fuels and chemicals in code-compliant, sturdy 18-gauge steel structures.
$2932.07 - $8121.88
Specifications
Certifications/Compliance | FM, NFPA, OSHA |
---|---|
Material | Steel (CR) |
Product Type | Can storage cabinet |
For Use With (Application) | Flammable liquid storage and protection |
Surface Coating | Epoxy, Polyester |
Catalog Number | Mfr. No. | Load Bearing Capacity | Color | Door Style | No. of Doors | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Load Bearing Capacity | Color | Door Style | No. of Doors | Price | Quantity | ||
17-985-57A
|
Justrite™
893080 |
30 gal. | Yellow | Self-close | 1 |
Each for $3,204.55
|
|
||
17-985-57B
|
Justrite™
894580 |
45 gal. | Yellow | Self-close | 1 |
Each for $8,121.88
|
|
||
18-955B
|
Justrite™
894503 |
45 gal. | Gray | Manual-close | 2 |
Each for $2,932.07
|
|
||
19-160-376
|
Justrite™
896025 |
60 gal. | White | Self-close | 2 |
Each for $3,983.06
|
|
||
19-120-2336
|
Justrite™
894520 |
45 gal. | Yellow | Self-close | 2 |
Each for $3,164.00
|
|
||
Description
Features of Justrite Flammables Cabinets
- Double-wall construction provides 1.5 in. of insulating air space to help prevent the spread of flames
- High-visibility Haz-Alert™ reflective warning labels offer added safety during power outages or fires
- Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away
- Easy-closing, self-latching doors facilitate fire protection of the cabinet
- Self latch engages critical three-point latching system for protection under fire conditions
- Fusible links hold self-closing doors wide open, flush with front of cabinet for easy access and resupply; automatically release and slam doors shut at 74°C (165°F)
- Concealed self-close mechanism provides optimum storage space and obstruction-free access to contents
- SpillSlope™ safety shelves direct incidental spills to the back and bottom of leakproof sump
- Adjustable leveling feet ensure solid positioning and alignment for tight door closure
- Exterior side panel has built-in grounding connector for easy grounding
- Finished inside and out with durable lead-free powder paint
- Lettering on door reads: “FLAMMABLE—KEEP FIRE AWAY”
- Accepts padlock (not included)
- Color: yellow, gray (optional), white (19-160-376)
Certifications
FM approved.
Compliance
Meet NFPA Code 30 and comply with OSHA regulations.
Warranty
10 years
Specifications
FM, NFPA, OSHA | |
Can storage cabinet | |
Epoxy, Polyester |
Steel (CR) | |
Flammable liquid storage and protection |