missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Justrite™ QuickBerm™ Decon Stations
Portable decon stations help prevent environmental contamination
$1704.14 - $3496.62
Catalog Number | Mfr. No. | Capacity (English) | Capacity (Metric) | Includes | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (English) | Capacity (Metric) | Includes | Price | Quantity | |||
19-111-075
|
Justrite™
28570 |
299 gal. | 1132 L | 2 Channels |
Each for $1,704.14
|
|
|||
19-111-076
|
Justrite™
28572 |
478 gal. | 1810 L | 3 Channels |
Each for $2,613.27
|
|
|||
19-111-081
|
Justrite™
28574 |
598 gal. | 2264 L | 4 Channels |
Each for $3,496.62
|
|
|||
Description
- Single-piece construction, inside wall-support system forms watertight wash down zones
- Quick and easy setup, no assembly required
- Available in two, three or four-zone models
- Watertight, lightweight PVC coated fabric provides wide range of chemical resistance
- The 203mm (8 in.) walls allow for easy entry and exit of compartments
- One 0.75 in. GHT drain fitting per zone
Specifications
Decon Station | |
PVC-coated Fabric |