Justrite™ Squeeze Bottle
Dispense a stream of flammable liquid directly onto a small component or subassembly. The squeeze bottle and stem are made from low-density polyethylene.
Supplier:  Justrite™ JMC14011
 View more versions of this product
Catalog No. 17-183-1
Explore available promotions
Description
Description
  • Airtight reduces spillage
  • Constructed of polyethylene with stainless-steel wire that allows stem to be shaped to reach hard-to-reach areas.
  • Bottle and stem are low-density polyethylene
Specifications
Specifications
946 mL (32 oz.)
32 oz.
3.5 in.
Polyethylene
232 mm
89 mm
946 mL
White
Stainless-steel wire within stem
Not Autoclavable
9.12 in.