Justrite™ Squeeze Bottle
Dispense a stream of flammable liquid directly onto a small component or subassembly. The squeeze bottle and stem are made from low-density polyethylene.
$85.62 - $87.83
Specifications
Color | White |
---|---|
Includes | Stainless-steel wire within stem |
Material | Polyethylene |
Autoclavable | Not Autoclavable |
Description
- Airtight reduces spillage
- Constructed of polyethylene with stainless-steel wire that allows stem to be shaped to reach hard-to-reach areas
- Bottle and stem are low-density polyethylene
Specifications
White | |
Stainless-steel wire within stem | |
Polyethylene | |
Not Autoclavable |