missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Justrite™ Sure-Grip™ EX Flammable Safety Cabinet
Sure-Grip™ EX Flammable Safety Cabinet protects workers, reduces fire risks, and improves worker productivity by allowing the storage of flammable fuels and chemicals in code-compliant, 18-gauge steel safety cabinets.
Supplier: Justrite™ 894500
Description
Sure-Grip™ EX Flammable Safety Cabinet protects workers, reduces fire risks, and improves worker productivity by allowing the storage of flammable fuels and chemicals in code-compliant, 18-gauge steel safety cabinets.
- Sure-Grip EX flush-mounted door handle does not interfere with traffic; comes with two keys for security and can be padlocked (padlock not included)
- Exclusive self-latching feature ensures that cabinet doors are safely closed without manually engaging door handle
- Double-wall construction provides 1.5 in. (3.8cm) of insulating air space to help prevent spread of flames High-visibility Haz-Alert™ reflective warning labels offer added safety during power outages or fires
- Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away Easy-closing, self-latching doors facilitate fire protection of the cabinet
- Self-latch engages critical three-point latching system for protection under fire conditions
- Fusible links hold self-closing doors wide open, flush with front of cabinet for easy access and resupply; automatically release and slam doors shut at 74°C (165°F)
- Concealed self-close mechanism provides optimum storage space and obstruction-free access to contents
- Two-door models hinged left and right have self-closing mechanism that ensures proper closing sequence regardless of which door is released first
- SpillSlope™ safety shelves direct incidental spills to the back and bottom of leakproof sump
- Adjustable leveling feet ensure solid positioning and alignment for tight door closure
- Exterior side panel has built-in grounding connector for easy grounding
- Finished inside and out with durable lead-free powder paint
- Meet OSHA and NFPA requirements
Certifications
FM approved except 19-120-2336.
Compliance
Meet NFPA Code 30 and OSHA regulations; models with self-close doors also comply with the Uniform Fire Code.
Warranty
10-Year
Specifications
100.3 cm | |
65 in. | |
Steel (CR) | |
108 kg | |
14.56 in. | |
45.7 cm | |
45 gal. | |
39.5 in. | |
153.7 cm | |
Flammable liquid storage and protection | |
109 cm | |
165.1 cm | |
2 |
37 cm | |
Epoxy, Polyester | |
U-Loc™ padlockable handle, Haz-Alert™ reflective trilingual warning labels, sturdy SpillSlope™ shelves, three-point Stainless Steel bullet self latching system and welded construction, dual vents with | |
18 in. | |
14.56 x 39.5 x 60.5 in. (37 x 100.3 x 153.7 cm) | |
237 lb. | |
2 | |
60.5 in. | |
Can storage cabinet | |
43 in. | |
Yellow | |
Manual-close | |
FM, NFPA, OSHA |