Learn More
Justrite™ Sure-Grip™ EX Flammable Safety Cabinet
Sure-Grip™ EX Flammable Safety Cabinet protects workers, reduces fire risks, and improves worker productivity by allowing the storage of flammable fuels and chemicals in code-compliant, 18-gauge steel safety cabinets.
$2049.68 - $8359.00
Specifications
Color | Yellow |
---|---|
Material | Steel (CR) |
Product Type | Can storage cabinet |
For Use With (Application) | Flammable liquid storage and protection |
Includes | U-Loc™ padlockable handle, Haz-Alert™ reflective trilingual warning labels, sturdy SpillSlope™ shelves, three-point Stainless Steel bullet self latching system and welded construction, dual vents with |
Catalog Number | Mfr. No. | Load Bearing Capacity | Dimensions (D x W x H) Interior | Door Style | Weight (Metric) | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Load Bearing Capacity | Dimensions (D x W x H) Interior | Door Style | Weight (Metric) | Price | Quantity | ||
19-120-2333
|
Justrite™
893000 |
30 gal. | 14.56 x 39.5 x 39.62 in. (37 x 100.3 x 100.6 cm) | Manual-close | 105 kg |
Each for $2,049.68
|
|
||
19-120-2334
|
Justrite™
893020 |
30 gal. | 14.56 x 39.5 x 39.62 in. (37 x 100.3 x 100.6 cm) | Self-close | 176 kg |
Each for $4,346.68
|
|
||
19-120-2335
|
Justrite™
894500 |
45 gal. | 14.56 x 39.5 x 60.5 in. (37 x 100.3 x 153.7 cm) | Manual-close | 108 kg |
Each for $2,683.21
|
|
||
19-120-2338
|
Justrite™
896020 |
60 gal. | 30.34 x 30.75 x 60.5 in. (77.1 x 78 x 153.7 cm) | Self-close | 218 kg |
Each for $6,687.21
|
|
||
19-120-2339
|
Justrite™
899000 |
90 gal. | 30.34 x 39.5 x 60.5 in. (77.1 x 100.3 x 153.7 cm) | Manual-close | 142 kg |
Each for $3,913.01
|
|
||
19-120-2340
|
Justrite™
899020 |
90 gal. | 30.34 x 39.5 x 60.5 in. (77.1 x 100.3 x 153.7 cm) | Self-close | 218 kg |
Each for $8,359.00
|
|
||
19-120-2337
|
Justrite™
896000 |
60 gal. | 30.34 x 30.75 x 60.5 in. (77.1 x 78 x 153.7 cm) | Manual-close | 141 kg |
Each for $5,650.71
|
|
||
Description
Sure-Grip™ EX Flammable Safety Cabinet protects workers, reduces fire risks, and improves worker productivity by allowing the storage of flammable fuels and chemicals in code-compliant, 18-gauge steel safety cabinets.
- Sure-Grip EX flush-mounted door handle does not interfere with traffic; comes with two keys for security and can be padlocked (padlock not included)
- Exclusive self-latching feature ensures that cabinet doors are safely closed without manually engaging door handle
- Double-wall construction provides 1.5 in. (3.8cm) of insulating air space to help prevent spread of flames High-visibility Haz-Alert™ reflective warning labels offer added safety during power outages or fires
- Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away Easy-closing, self-latching doors facilitate fire protection of the cabinet
- Self-latch engages critical three-point latching system for protection under fire conditions
- Fusible links hold self-closing doors wide open, flush with front of cabinet for easy access and resupply; automatically release and slam doors shut at 74°C (165°F)
- Concealed self-close mechanism provides optimum storage space and obstruction-free access to contents
- Two-door models hinged left and right have self-closing mechanism that ensures proper closing sequence regardless of which door is released first
- SpillSlope™ safety shelves direct incidental spills to the back and bottom of leakproof sump
- Adjustable leveling feet ensure solid positioning and alignment for tight door closure
- Exterior side panel has built-in grounding connector for easy grounding
- Finished inside and out with durable lead-free powder paint
- Meet OSHA and NFPA requirements
Certifications
FM approved except 19-120-2336.
Compliance
Meet NFPA Code 30 and OSHA regulations; models with self-close doors also comply with the Uniform Fire Code.
Warranty
10-Year
Specifications
Yellow | |
Can storage cabinet | |
U-Loc™ padlockable handle, Haz-Alert™ reflective trilingual warning labels, sturdy SpillSlope™ shelves, three-point Stainless Steel bullet self latching system and welded construction, dual vents with | |
Epoxy, Polyester |
Steel (CR) | |
Flammable liquid storage and protection | |
2 |