Learn More
Kappler™ Chem-Tape™
Seals gaps in protective apparel at sleeve, ankle, hood and storm flap
Supplier:  Kappler™ 99402YW1
 View more versions of this product
Catalog No. 19-036-34
Only null left
Explore available promotions
Description
Description
Seals gaps in protective apparel at sleeve, ankle, hood and storm flap
Yellow adhesive tape attaches pieces of protective apparel ensemble, preventing gross liquid intrusion at interface points.
  • Provides a durable reinforcement, but tears easily for quick application
  • For single use only
Order Info
Roll measures 2 in. x 60 yd. (5cm x 55m).
Compliance
Meets ASTM™ F739 (using ASTM F1001 test battery) standards; permeation tested in accordance with MIL-STD 282 (using various chemical warfare agents).
Specifications
Specifications
Chem-tape
Single-use
Chemicals, Liquid
Chem-tape
Ideal to attach pieces of protective apparel ensemble, prevents gross liquid intrusion into gaps at interface points
Yellow
Hooded
ASTM F739, MIL-STD 282
2 in. x 60 yd.
Product Suggestions
Product Suggestions