missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Kimberly-Clark Professional™ KleenGuard™ KGA10 Lab Coats
Highly breathable spunbond fabric lab coats are CE Category I PPE and are free from natural rubber latex and silicone materials.
Supplier: Kimberly-Clark Professional™ 67318
Description
- For non-hazardous particulate protection
- Lightweight and highly breathable spunbond fabric allows moisture to escape for maximum comfort
- 4-Snap Closure, Elastic Wrists, No Pockets
- Not made with natural rubber latex or silicone
- Recyclable with RightCycle™ by Kimberly-Clark Professional™
Specifications
General Purpose Lab Coat | |
Unisex | |
Snaps | |
Single-use | |
None | |
KGA10 |
3X-Large | |
White | |
Elastic | |
Non-sterile | |
Yes | |
Stitched |