missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Kimberly-Clark Professional™ KleenGuard™ KGA10 Lab Coats
Highly breathable spunbond fabric lab coats are CE Category I PPE and are free from natural rubber latex and silicone materials.
$143.72 - $149.47
Specifications
Product Type | General Purpose Lab Coat |
---|---|
Gender | Unisex |
Color | White |
Closure Type | Snaps |
Cuff Style | Elastic |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
17-101-157
|
Kimberly-Clark Professional™
67314 |
Medium |
Case of 50 Each for $143.72
|
|
|||||
17-101-158
|
Kimberly-Clark Professional™
67315 |
Large |
Case of 50 Each for $143.72
|
|
|||||
17-101-159
|
Kimberly-Clark Professional™
KC67316 |
X-Large |
Case of 50 Each for $143.72
|
|
|||||
17-101-160
|
Kimberly-Clark Professional™
67317 |
2X-Large |
Case of 50 Each for $143.72
|
|
|||||
17-101-161
|
Kimberly-Clark Professional™
67318 |
3X-Large |
Case of 50 Each for $149.47
|
|
|||||
Description
- For non-hazardous particulate protection
- Lightweight and highly breathable spunbond fabric allows moisture to escape for maximum comfort
- 4-Snap Closure, Elastic Wrists, No Pockets
- Not made with natural rubber latex or silicone
- Recyclable with RightCycle™ by Kimberly-Clark Professional™
Specifications
General Purpose Lab Coat | |
White | |
Elastic | |
Non-sterile | |
Yes | |
Stitched |
Unisex | |
Snaps | |
Single-use | |
None | |
KGA10 |