DWK Life Sciences Kimble™ KIMAX™ Burets with 3-Way PTFE Stopcock
Class A
Supplier:  DWK Life Sciences 17052F25
 View more versions of this product
Catalog No. 03-724-10A
Explore available promotions
Includes: No. 2 PTFE stopcock and KimKap™ dust cap.
Description
Description
Class A
  • Precision bore tubing
  • Durable white scale
  • Permanently marked with individual serial number
  • Filling tube accepts 1/4 in. I.D. flexible tubing
Certifications
A certificate is provided to indicate instrument traceability to standards provided by NIST™.
Compliance
Meet ASTM™ E 287
Specifications
Specifications
25 mL
Borosilicate Glass
General Purpose Titrations Requiring Traceable Volumetric Accuracy
Dust Cap
PTFE
Automatic Buret
ASTM E287, ASTM E438, ASTM E542, ASTM E694, NIST
617 mm
0.1 mL
24.29 in.
White
±0.03 mL
Class A