missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Kimble™ KIMAX™ Burets with Threaded Closure Fit

Educational grade

$347.38 - $362.06

Specifications

Material Borosilicate Glass
Graduations 0.1 mL
For Use With (Application) Educational Institutions and for Many General Laboratory Procedures
Includes Dust Cap
Scale Black
View More Specs

Includes: KimKap™ dust cap.

Products 2
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
03-857A
View Documents Promotion Details
DWK Life Sciences
17121H25
25 mL
Case for $347.38
 
03-857B
View Documents Promotion Details
DWK Life Sciences
17121H50
50 mL
Case for $362.06
 
Description

Description

 

  • Economical and versatile
  • Accept PTFE stopcock assembly with threaded closure to prevent dislodging
  • Tube has a reinforced top with tooled lower end
  • Ground tip fits into lower end of stopcock assembly
  • Quantity: 1/Cs.

 

Specifications

Specifications

Borosilicate Glass
Educational Institutions and for Many General Laboratory Procedures
Black
Educational Grade Buret
ASTM E438
0.1 mL
Dust Cap
PTFE
Class A
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'