missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits
DWK Life Sciences Kimble™ Reusable Class A Volumetric Serialized and Certified Pipets, TD
Designed from ASTM Specification E969, Class A requirements. Kimble™ Reusable Class A Volumetric Serialized and Certified Pipets, TD are ideal for measuring accurate volumes of liquids.
$292.59 - $562.68
Spécifications
Autoclavable | Autoclavable |
---|---|
Class | A |
Material | Borosilicate Glass |
Type | Serialized Volumetric Pipet |
Disposable | Reusable |
Numéro de catalogue | Numéro du manufacturier. | Volume (Metric) | Length (Metric) | Tolerance | Prix | Quantité | |||
---|---|---|---|---|---|---|---|---|---|
Numéro de catalogue | Numéro du manufacturier. | Volume (Metric) | Length (Metric) | Tolerance | Prix | Quantité | |||
02-555-344
|
DWK Life Sciences
37006510 |
0.5 mL | 31 cm | ±0.006 mL |
caisse de 6 chaque for $318.47
|
|
|||
02-555-345
|
DWK Life Sciences
370061 |
1 mL | 31 cm | ±0.006 mL |
caisse de 6 chaque for $319.79
|
|
|||
02-555-347
|
DWK Life Sciences
370062 |
2 mL | 33.5 cm | ±0.006 mL |
caisse de 6 chaque for $416.35
|
|
|||
02-555-346
|
DWK Life Sciences
370061510 |
1.5 mL | 33.5 cm | ±0.006 mL |
caisse de 6 chaque for $380.94
|
|
|||
02-555-348
|
DWK Life Sciences
370062510 |
2.5 mL | 35 cm | ±0.006 mL |
caisse de 6 chaque for $432.00
|
|
|||
02-555-349
|
DWK Life Sciences
370063 |
3 mL | 35 cm | ±0.01 mL |
caisse de 6 chaque for $472.74
|
|
|||
02-555-350
|
DWK Life Sciences
370064 |
4 mL | 35 cm | ±0.01 mL |
caisse de 6 chaque for $520.50
|
|
|||
02-555-351
|
DWK Life Sciences
370065 |
5 mL | 36.5 cm | ±0.01 mL |
caisse de 6 chaque for $528.47
|
|
|||
02-555-352
|
DWK Life Sciences
370066 |
6 mL | 37.5 cm | ±0.01 mL |
caisse de 6 chaque for $538.50
|
|
|||
02-555-353
|
DWK Life Sciences
370067 |
7 mL | 40 cm | ±0.01 mL |
caisse de 6 chaque for $547.00
|
|
|||
02-555-354
|
DWK Life Sciences
370068 |
8 mL | 40 cm | ±0.01 mL |
caisse de 6 chaque for $554.62
|
|
|||
02-555-355
|
DWK Life Sciences
370069 |
9 mL | 40 cm | ±0.02 mL |
caisse de 6 chaque for $562.68
|
|
|||
02-555-356
|
DWK Life Sciences
3700610 |
10 mL | 40 cm | ±0.02 mL |
caisse de 6 chaque for $558.50
|
|
|||
02-555-357
|
DWK Life Sciences
3700612 |
12 mL | 46 cm | ±0.02 mL |
caisse de 3 chaque for $292.59
|
|
|||
02-555-358
|
DWK Life Sciences
3700615 |
15 mL | 45.5 cm | ±0.03 mL |
caisse de 3 chaque for $314.38
|
|
|||
02-555-359
|
DWK Life Sciences
3700620 |
20 mL | 50.5 cm | ±0.03 mL |
caisse de 3 chaque for $354.56
|
|
|||
02-555-360
|
DWK Life Sciences
3700625 |
25 mL | 50.5 cm | ±0.03 mL |
caisse de 3 chaque for $359.85
|
|
|||
02-555-361
|
DWK Life Sciences
3700630 |
30 mL | 52.5 cm | ±0.03 mL |
caisse de 3 chaque for $365.50
|
|
|||
02-555-362
|
DWK Life Sciences
3700640 |
40 mL | 52.5 cm | ±0.03 mL |
caisse de 3 chaque for $408.32
|
|
|||
02-555-363
|
DWK Life Sciences
3700650 |
50 mL | 54.5 cm | ±0.05 mL |
caisse de 3 chaque for $421.38
|
|
|||
02-555-365
|
DWK Life Sciences
37006100 |
100 mL | 55 cm | ±0.08 mL |
caisse de 3 chaque for $509.91
|
|
|||
02-555-364
|
DWK Life Sciences
3700675 |
75 mL | 55 cm | ±0.05 mL |
caisse de 3 chaque for $487.24
|
|
|||
Description
Designed from ASTM Specification E969, Class A requirements. Kimble™ KIMAX™ Reusable Class A Volumetric Serialized Pipets, TD are ideal for measuring accurate volumes of liquids.
- Includes individual Certificates of Calibration
- Sizes 1mL and larger are marked with large numerals on the bulb, indicating capacity
- Color-coded (ASTM E1273) for ease in sorting and selecting the correct pipet
- With a legend of permanent brown stain
- Permanently marked individual serial number
- Autoclavable
- 75 mL size designed from ASTM E542
- Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Spécifications
Autoclavable | |
Borosilicate Glass | |
Reusable |
A | |
Serialized Volumetric Pipet |