missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimberly-Clark Professional™ KleenGuard™ G80 Nitrile Chemical Resistant Gloves

Green, hand-specific gloves provide maximum protection against chemical and mechanical hazards. KleenGuard™ G80 Nitrile Chemical Resistant Gloves include improved, more consistent cotton flock lining for added comfort.

Supplier:  Kimberly-Clark Professional™ 94445

 View more versions of this product

Catalog No. 19-812-419


Explore available promotions

Description

Description

  • Hand specific latex-free gloves protect hands against chemical or mechanical hazards
  • Diamond embossed fingertips offer enhanced wet and dry grip
  • Flock-lined for easy donning and extra comfort in extended use
  • Improved AQL of 0.65 versus 1.5, resulting in fewer pinholes
  • Modified Nitrile formulation for more durability and enhanced chemical resistance.
  • Now available in 5 bulk-packed poly bags versus individually wrapped pairs
  • Ideal for: providing PPE category 3 protection, level 4 abrasion resistance and durability in manufacturing, transport, construction, agriculture cleaning and janitorial work, when handling chemicals, oils, grease, acids, caustics or solvents
Specifications

Specifications

FDA, ASTM D3767, ASTM D412, ASTM D5151, ASTM D6319
No
15 mil
Yes
15.7 mil
Nitrile Gloves
15 mil
EN 388:4101X
Type A
Bacteria, Fungi only
Rough
Lined
KleenGuard™ G80
33 cm
CE 0120, EN 374-3
Green
0.38 mm
Nitrile
0.4 mm
Small
0.38 mm
No
JKLNOP
Acids, bases, Solvents and Caustics
Cotton Flock
94445
13 in.
Videos
SDS
Documents

Documents

Promotions

Promotions