missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Kimberly-Clark Professional™ KleenGuard™ G80 Nitrile Chemical Resistant Gloves
Green, hand-specific gloves provide maximum protection against chemical and mechanical hazards. KleenGuard™ G80 Nitrile Chemical Resistant Gloves include improved, more consistent cotton flock lining for added comfort.
$254.27 - $289.61
Specifications
Certifications/Compliance | FDA, ASTM D3767, ASTM D412, ASTM D5151, ASTM D6319 |
---|---|
Chlorinated | No |
Color | Green |
Finger Thickness (English) | 15 mil |
Finger Thickness (Metric) | 0.38 mm |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
19-812-419
|
Kimberly-Clark Professional™
94445 |
Small |
Case of 60 Each for $280.20
|
|
|||||
19-812-420
|
Kimberly-Clark Professional™
94446 |
Medium |
Case of 60 Each for $254.27
|
|
|||||
19-812-421
|
Kimberly-Clark Professional™
94447 |
Large |
Case of 60 Each for $289.61
|
|
|||||
19-812-422
|
Kimberly-Clark Professional™
94448 |
X-Large |
Case of 60 Each for $280.20
|
|
|||||
19-812-423
|
Kimberly-Clark Professional™
94449 |
2X-Large |
Case of 60 Each for $280.20
|
|
|||||
Description
- Hand specific latex-free gloves protect hands against chemical or mechanical hazards
- Diamond embossed fingertips offer enhanced wet and dry grip
- Flock-lined for easy donning and extra comfort in extended use
- Improved AQL of 0.65 versus 1.5, resulting in fewer pinholes
- Modified Nitrile formulation for more durability and enhanced chemical resistance.
- Now available in 5 bulk-packed poly bags versus individually wrapped pairs
- Ideal for: providing PPE category 3 protection, level 4 abrasion resistance and durability in manufacturing, transport, construction, agriculture cleaning and janitorial work, when handling chemicals, oils, grease, acids, caustics or solvents
Specifications
FDA, ASTM D3767, ASTM D412, ASTM D5151, ASTM D6319 | |
Green | |
0.38 mm | |
Nitrile | |
0.4 mm | |
15 mil | |
EN 388:4101X | |
Type A | |
Bacteria, Fungi only | |
Rough | |
Lined | |
13 in. |
No | |
15 mil | |
Yes | |
15.7 mil | |
Nitrile Gloves | |
0.38 mm | |
No | |
JKLNOP | |
Acids, bases, Solvents and Caustics | |
Cotton Flock | |
KleenGuard™ G80 | |
33 cm |