Kimberly-Clark Professional™ KleenGuard™ V50 Epic™ Safety Eyewear
Kimberly-Clark™ Professional KleenGuard™ V50 Epic™ Safety Eyewear feature a flexible foam gasket, which provides long term comfort and helps to seal out dust and debris.
Supplier:  Kimberly-Clark Professional™ 33345
 View more versions of this product
Catalog No. 19-006-208
Explore available promotions
Description
Description
  • Anti-fogging solution resists fogging up to two times longer than the EN166 industry standard
  • Direct ventilation at the top and bottom of the eyewear to provide superior anti-fog performance for worker efficiency and safety
  • Meets ANSI Z87.1+ standards and CSA Z94.3 standards
  • Lenses are ideal for indoor environments, thanks to their maximum light transmission
  • Temple bars can be interchanged with an included head strap when a closer, more secure fit is required
  • Wrap around design and impact resistant lenses make these ideal for use in dirty/metal manufacturing, transportation, as motorcycle goggles. etc.
Specifications
Specifications
Clear Super
Safety Eyewear
Red
Clear
Black
Polycarbonate