Kimberly-Clark Professional™ KleenGuard™ V50 Epic™ Safety Eyewear
Kimberly-Clark™ Professional KleenGuard™ V50 Epic™ Safety Eyewear feature a flexible foam gasket, which provides long term comfort and helps to seal out dust and debris.
Supplier: Kimberly-Clark Professional™ 33347
Description
- Anti-fogging solution resists fogging up to two times longer than the EN166 industry standard
- Direct ventilation at the top and bottom of the eyewear to provide superior anti-fog performance for worker efficiency and safety
- Meets ANSI Z87.1+ standards and CSA Z94.3 standards
- Lenses are ideal for indoor environments, thanks to their maximum light transmission
- Temple bars can be interchanged with an included head strap when a closer, more secure fit is required
- Wrap around design and impact resistant lenses make these ideal for use in dirty/metal manufacturing, transportation, as motorcycle goggles. etc.
Specifications
Indoor/Outdoor Super | |
Safety Eyewear | |
Red |
Indoor/Outdoor | |
Black | |
Polycarbonate |