Kimberly-Clark Professional™ KleenGuard™ V50 OTG™ Safety Glasses
When you need to wear your prescription glasses and safety eye protection, you can trust KleenGuard OTG Over the Glasses Safety Glasses. They are made with a nylon frame for strength, durability and comfort and fit over most prescription glasses.
Supplier:  Kimberly-Clark Professional™ 20747
 View more versions of this product
Catalog No. 29-636-6553
Explore available promotions
Description
Description
  • Made with a nylon frame for strength, durability and comfort and fit over most prescription glasses
  • See-through integrated side shields provide side protection without hindering peripheral vision
  • Hard coated for scratch resistance and longer life
  • Anti-fog coating for humid environment and temperature variances
  • Meet ANSI Z87.1+ standards for high impact
  • Polycarbonate lenses provide 99.9% UVA / UVB / UVC protection
Specifications
Specifications
ANSI Z87.1 and CSA Z94.3
OTG safety spectacles
Polycarbonate
UV400
Smoke
Black
Polycarbonate