Kimberly-Clark Professional™ KleenGuard™ V50 OTG™ Safety Glasses
When you need to wear your prescription glasses and safety eye protection, you can trust KleenGuard OTG Over the Glasses Safety Glasses. They are made with a nylon frame for strength, durability and comfort and fit over most prescription glasses.
$9.06 - $109.65
Specifications
Certifications/Compliance ANSI Z87.1 and CSA Z94.3
Product Type OTG safety spectacles
Frame Color Black
Material Polycarbonate
Frame Material Polycarbonate
View More Specs
Products 3
Catalog Number Mfr. No. Lens Tint Price Quantity  
Catalog Number Mfr. No. Lens Tint Price Quantity  
19-818754
View Documents
Kimberly-Clark Professional™
20746
Clear
Each for $9.06
Case of 12 Each for $105.36
29-636-6553
View Documents
Kimberly-Clark Professional™
20747
Smoke
Case of 12 Each for $109.65
29-636-6554
View Documents
Kimberly-Clark Professional™
20748
Indoor/Outdoor, Indoor/Outdoor Mirror
Case of 12 Each for $109.65
Description
Description
  • Made with a nylon frame for strength, durability and comfort and fit over most prescription glasses
  • See-through integrated side shields provide side protection without hindering peripheral vision
  • Hard coated for scratch resistance and longer life
  • Anti-fog coating for humid environment and temperature variances
  • Meet ANSI Z87.1+ standards for high impact
  • Polycarbonate lenses provide 99.9% UVA / UVB / UVC protection
Specifications
Specifications
ANSI Z87.1 and CSA Z94.3
Black
Polycarbonate
OTG safety spectacles
Polycarbonate
UV400
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'