Kimberly-Clark Professional™ KleenGuard™ V50 OTG™ Safety Glasses
When you need to wear your prescription glasses and safety eye protection, you can trust KleenGuard OTG Over the Glasses Safety Glasses. They are made with a nylon frame for strength, durability and comfort and fit over most prescription glasses.
Supplier: Kimberly-Clark Professional™ 20748
Description
- Made with a nylon frame for strength, durability and comfort and fit over most prescription glasses
- See-through integrated side shields provide side protection without hindering peripheral vision
- Hard coated for scratch resistance and longer life
- Anti-fog coating for humid environment and temperature variances
- Meet ANSI Z87.1+ standards for high impact
- Polycarbonate lenses provide 99.9% UVA / UVB / UVC protection
Specifications
UV-treated | |
Indoor/Outdoor, Indoor/Outdoor Mirror | |
Black | |
Polycarbonate |
ANSI Z87.1 and CSA Z94.3 | |
OTG safety spectacles | |
Polycarbonate | |
UV400 |