Kimberly-Clark Professional™ Nemesis™ Rx Reader Safety Glasses
Kimberly-Clark™ Professional Nemesis™ Rx Reader Safety Glasses are designed for long-term comfort, thanks to flexible and lightweight design, soft touch temples and a nose piece with integrated ridges to channel away sweat
Supplier: Kimberly-Clark Professional™ 28618
Description
- Equipped to suit those who need vision assistance
- Polycarbonate lenses provide 99.9% UVA/ UVB/UVC protection to help prevent eye damage like cataracts, retinal damage, and other conditions that can cause temporary vision loss
- Wraparound frame has a base curve of 8 that extends the range of peripheral protection, as well as soft touch temples for comfort
- Corrective lenses are hard coated for scratch resistance and longer life
- Each pair comes with a neck cord for securing
- Manufactured to comply with the ANSI / ISEA Z87.1+ standard for personal eye protection
- Comfortable and functional for construction, manufacturing, and a host of other industries
- Patented FLEX-DRY designed to be extremely flexible, to channel sweat away from the eyes, and to help prevent slippage down the nose for long-term comfort
Certifications
Meets ANSI Z87.1-2010 Impact standards
CE EN 166: 2001 (Clear lens only); CSA Z94.3-07 (Clear lens only).
Specifications
ANSI Z87.1; CE EN 166: 2001;CSA Z94.3-07 | |
Reader Safety Glasses | |
Black | |
Polycarbonate | |
UV400 |
Clear | |
Industrial manufacturing, outdoor environments, impact, splash, grinding/chipping, shaving/sanding | |
+1.00 | |
Nylon |