Kimberly-Clark Professional™ KleenGuard™ V60 30-06™ Readers Safety Eyewear
KleenGuard V60 30-06 Readers Safety Glasses are equipped to suit those who need vision assistance and eye protection. Available in a variety of lens and frame colors and prescription strengths, they are designed for long-term comfort.
Supplier:  Kimberly-Clark Professional™ 19878
 View more versions of this product
Catalog No. 19-130-0919
Explore available promotions
Description
Description
  • Stylish ergonomic black nylon frame
  • Sleek seven-base clear polycarbonate lenses
  • Soft rubber nose pad with self-adjusting fingers
  • Magnification insert is molded into the lens
  • UV protection up to 400 nanometers
Compliance
Exceeds ANSI Z87.1-2010 High Velocity Industrial Protection Standards.
Specifications
Specifications
ANSI Z87.1-2010
Prescription Safety Glasses
+1.50
Nylon
Clear
Black
Polycarbonate
UV400