missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Kimberly-Clark Professional™ KleenGuard™ V60 30-06™ Readers Safety Eyewear
KleenGuard V60 30-06 Readers Safety Glasses are equipped to suit those who need vision assistance and eye protection. Available in a variety of lens and frame colors and prescription strengths, they are designed for long-term comfort.
$15.96 - $15.96
Specifications
Certifications/Compliance | ANSI Z87.1-2010 |
---|---|
Lens Tint | Clear |
Frame Color | Black |
Lens Material | Polycarbonate |
Frame Material | Nylon |
Catalog Number | Mfr. No. | Magnification Power | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Magnification Power | Price | Quantity | |||||
19-130-0919
|
Kimberly-Clark Professional™
19878 |
+1.50 |
Each for $15.96
Case of 6 Each for $95.76
|
|
|||||
Description
- Stylish ergonomic black nylon frame
- Sleek seven-base clear polycarbonate lenses
- Soft rubber nose pad with self-adjusting fingers
- Magnification insert is molded into the lens
- UV protection up to 400 nanometers
Compliance
Exceeds ANSI Z87.1-2010 High Velocity Industrial Protection Standards.
Specifications
ANSI Z87.1-2010 | |
Black | |
Nylon |
Clear | |
Polycarbonate | |
UV400 |