Kimberly-Clark Professional™ Kimtech Prep™ Kimtex™ Wipers
Heavy-duty wiper for oil and solvent cleanups
Supplier:  Kimberly-Clark Professional™ 33560
 View more versions of this product
Catalog No. 15-235-85
Explore available promotions
Description
Description
  • Reusable wiper is excellent for cleaning and prep on surfaces and parts and for glass cleaning
  • Constructed of low-lint polypropylene for long-lasting performance
  • Absorb liquids quickly — ideal for big cleanups
  • Acid-, base-, and solvent-resistant
  • Blue
Specifications
Specifications
Heavy-duty Wiper
33 cm
31.75 cm
Blue
13 x 12.5 in. (33 x 31.75 cm)
13 in.
12 in.
Polypropylene
Reusable
General-Purpose
Product Suggestions
Product Suggestions