missing translation for 'onlineSavingsMsg'
Learn More

Ansell Kimtech™ A5 Cleanroom Apparel, Bulk Packaging

Coveralls feature a polypropylene breathable SMS fabric with a cloth-like feel.

$269.94 - $304.18

Specifications

Product Type Cleanroom Coveralls
Color White
Disposable Single-use
Cuff Style Thumb Loop
Neck Style Collared
View More Specs

Products 9
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-169-052
View Documents
Ansell
49831
Small
Case of 25 Each for $279.59
Only null left
 
19-169-053
View Documents
Ansell
49832
Medium
Case of 25 Each for $270.56
Only null left
 
19-169-054
View Documents
Ansell
49833
Large
Case of 25 Each for $270.56
Only null left
 
19-169-055
View Documents
Ansell
49834
X-Large
Case of 25 Each for $270.56
Only null left
 
19-169-056
View Documents
Ansell
49835
2X-Large
Case of 25 Each for $280.80
Only null left
 
19-169-057
View Documents
Ansell
49836
3X-Large
Case of 25 Each for $292.31
Only null left
 
19-169-058
View Documents
Ansell
49837
4X-Large
Case of 25 Each for $269.94
Only null left
 
19-169-059
View Documents
Ansell
49838
5X-Large
Case of 25 Each for $292.36
Only null left
 
19-169-060
View Documents
Ansell
49841
6X-Large, 7X-Large, 8X-Large
Case of 25 Each for $304.18
Only null left
 
Description

Description

Kimtech™ A5 Cleanroom Coveralls are made of 100% polypropylene breathable SMS fabric, which gives the wearer cloth-like comfort. While other coveralls for cleanroom environments may look similar, the ones from Kimtech™ Pure have distinct advantages including tightly bound seams to strengthen the barrier, a high neck collar to reduce contamination, thumb loops to keep the sleeves from rolling back and a zipper flap to cover the zipper area.

In addition, this protective apparel is recyclable through the RightCycle program.

This personal protective apparel is triple-bagged and vacuum packaged, suitable for ISO Class 5 or higher cleanrooms. It is not made with natural rubber latex, silicone, or the BHT preservative and is designed for single-use (disposable PPE).

For use in biotech, medical device, electronics and telecommunications manufacturing, they are designed to protect your process by providing comfortable, sustainable protection.

  • Made of breathable SMS fabric, the coveralls feature a high-neck collar, a zipper flap, thumb loops and bound seams
  • Triple bagged, vacuum packed, suitable for ISO Class 5 or higher cleanrooms; CE certified
  • Kimtech™ A5 Cleanroom Apparel is manufactured in an ISO-certified facility, clean-processes and individually packaged under cleanroom conditions
  • Recyclable through the RightCycle program, which can help you meet your sustainability goals
Specifications

Specifications

Cleanroom Coveralls
Single-use
Collared
Standing
Class 100-100,000 (ISO 5-8)
White
Thumb Loop
Non-sterile
Compounding, Mixing, Filling, Cleaning, Pharmaceutical, Biomedial Research
Videos
SDS
Documents

Documents