Kimberly-Clark Professional™ KleenGuard™ A10 Light-Duty Bouffant Caps
Lightweight spunbound material provides disposable protection against dirt and grime.
Supplier:  Kimberly-Clark Professional™ 36860
 View more versions of this product
Catalog No. 17-981-43B
Explore available promotions
Description
Description
Lightweight spunbound material provides disposable protection against dirt and grime.
  • Single layer, spunbond polypropylene
  • Latex-free
  • White
Order Info
Available in case of 1000 caps (in 10 polybags containing 100 caps each).
Specifications
Specifications
Bouffant
Spunbond Polypropylene
Yes
Food Processing, General Maintenance, and Manufacturing
24 in.
Single-use
36860