Kimberly-Clark Professional™ KleenGuard™ A70 Chemical Spray Coveralls
Reflex™ design offers 12% more chest room and 6% longer body length
Supplier:  Kimberly-Clark Professional™ 09813
 View more versions of this product
Catalog No. 29-635-8884
Explore available promotions
Description
Description
Coveralls provide protection for hazardous waste remediation, environmental cleanup, petrochemical and oil refining, acid and caustic handling, biohazard cleanup, tank cleaning and pharmaceutical manufacturing.
  • Feature bound seams
  • Storm flap provides a seamless chest area
  • Water-resistant zipper and wick-away lining for enhanced comfort
  • Yellow
Compliance
Meet ASTM™ F 903 penetration test standards.
Specifications
Specifications
Large
Yellow
Elastic
Front Zipper with Storm Flap
Chemicals
Zipper Front, Storm Flap, Elastic Wrists, Ankles and Hood
Hazardous waste remediation, environmental cleanup, petrochemical refining, Acid/Caustic handling, biohazard cleanup, tank cleaning, pharmaceutical manufacturing
Polyethylene, Spunbond Polypropylene
Elastic
Hooded
Bound
ASTM F 903, NFPA 99
1.5 mil