missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Kimberly-Clark Professional™ KleenGuard™ A70 Chemical Spray Coveralls
Reflex™ design offers 12% more chest room and 6% longer body length
$187.70 - $252.16
Specifications
Material | Polyethylene, Spunbond Polypropylene |
---|---|
Color | Yellow |
Ankle Style | Elastic |
Cuff Style | Elastic |
Neck Style | Hooded |
Catalog Number | Mfr. No. | Description | Size | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Size | Price | Quantity | ||||
19-166-488
|
Kimberly-Clark Professional™
00685 |
Zipper Front, Storm Flap, Elastic Wrists, Ankles, Hood and Boots | 2X-Large |
Case of 12 Each for $234.20
|
|
||||
19-166-487
|
Kimberly-Clark Professional™
00684 |
Zipper Front, Storm Flap, Elastic Wrists, Ankles, Hood and Boots | X-Large |
Case of 12 Each for $221.55
|
|
||||
29-635-8893
|
Kimberly-Clark Professional™
00686 |
Zipper Front, Storm Flap, Elastic Wrists, Ankles, Hood and Boots | 3X-Large |
Case of 12 Each for $252.16
|
|
||||
29-635-8894
|
Kimberly-Clark Professional™
00687 |
Zipper Front, Storm Flap, Elastic Wrists, Ankles, Hood and Boots | 4X-Large |
Case of 12 Each for $246.65
|
|
||||
29-635-8883
|
Kimberly-Clark Professional™
09812 |
Zipper Front, Storm Flap, Elastic Wrists, Ankles and Hood | Medium |
Case of 12 Each for $197.55
|
|
||||
29-635-8884
|
Kimberly-Clark Professional™
09813 |
Zipper Front, Storm Flap, Elastic Wrists, Ankles and Hood | Large |
Case of 12 Each for $187.70
|
|
||||
29-635-8885
|
Kimberly-Clark Professional™
09814 |
Zipper Front, Storm Flap, Elastic Wrists, Ankles and Hood | X-Large |
Case of 12 Each for $199.24
|
|
||||
29-635-8888
|
Kimberly-Clark Professional™
09817 |
Zipper Front, Storm Flap, Elastic Wrists, Ankles and Hood | 4X-Large |
Case of 12 Each for $221.36
|
|
||||
29-635-8886
|
Kimberly-Clark Professional™
09815 |
Zipper Front, Storm Flap, Elastic Wrists, Ankles and Hood | 2X-Large |
Case of 12 Each for $208.14
|
|
||||
19-166-495
|
Kimberly-Clark Professional™
09816 |
Zipper Front, Storm Flap, Elastic Wrists, Ankles and Hood | 3X-Large |
Case of 12 Each for $220.23
|
|
||||
19-812-900
|
Kimberly-Clark Professional™
09818 |
Zipper Front, Storm Flap, Elastic Wrists, Ankles and Hood | 5X-Large |
Case of 12 Each for $222.31
|
|
||||
29-635-8890
|
Kimberly-Clark Professional™
00683 |
Zipper Front, Storm Flap, Elastic Wrists, Ankles, Hood and Boots | Large |
Case of 12 Each for $242.32
|
|
||||
Description
Coveralls provide protection for hazardous waste remediation, environmental cleanup, petrochemical and oil refining, acid and caustic handling, biohazard cleanup, tank cleaning and pharmaceutical manufacturing.
- Feature bound seams
- Storm flap provides a seamless chest area
- Water-resistant zipper and wick-away lining for enhanced comfort
- Yellow
Compliance
Meet ASTM™ F 903 penetration test standards.
Specifications
Polyethylene, Spunbond Polypropylene | |
Elastic | |
Hooded | |
Bound | |
ASTM F 903, NFPA 99 | |
Hazardous waste remediation, environmental cleanup, petrochemical refining, Acid/Caustic handling, biohazard cleanup, tank cleaning, pharmaceutical manufacturing |
Yellow | |
Elastic | |
Front Zipper with Storm Flap | |
Chemicals | |
1.5 mil |