Kimberly-Clark Professional™ KleenGuard™ A70 Chemical Spray Coveralls
Reflex™ design offers 12% more chest room and 6% longer body length
Supplier:  Kimberly-Clark Professional™ 09817
 View more versions of this product
Catalog No. 29-635-8888
Explore available promotions
Description
Description
Coveralls provide protection for hazardous waste remediation, environmental cleanup, petrochemical and oil refining, acid and caustic handling, biohazard cleanup, tank cleaning and pharmaceutical manufacturing.
  • Feature bound seams
  • Storm flap provides a seamless chest area
  • Water-resistant zipper and wick-away lining for enhanced comfort
  • Yellow
Compliance
Meet ASTM™ F 903 penetration test standards.
Specifications
Specifications
4X-Large
Yellow
Elastic
Front Zipper with Storm Flap
Chemicals
Zipper Front, Storm Flap, Elastic Wrists, Ankles and Hood
Hazardous waste remediation, environmental cleanup, petrochemical refining, Acid/Caustic handling, biohazard cleanup, tank cleaning, pharmaceutical manufacturing
Polyethylene, Spunbond Polypropylene
Elastic
Hooded
Bound
ASTM F 903, NFPA 99
1.5 mil