missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Kimberly-Clark Professional™ KleenGuard™ A70 Chemical Spray Coveralls
Reflex™ design offers 12% more chest room and 6% longer body length
Supplier: Kimberly-Clark Professional™ 00686
Description
Coveralls provide protection for hazardous waste remediation, environmental cleanup, petrochemical and oil refining, acid and caustic handling, biohazard cleanup, tank cleaning and pharmaceutical manufacturing.
- Feature bound seams
- Storm flap provides a seamless chest area
- Water-resistant zipper and wick-away lining for enhanced comfort
- Yellow
Compliance
Meet ASTM™ F 903 penetration test standards.
Specifications
3X-Large | |
Yellow | |
Elastic | |
Front Zipper with Storm Flap | |
Chemicals | |
Zipper Front, Storm Flap, Elastic Wrists, Ankles, Hood and Boots | |
Hazardous waste remediation, environmental cleanup, petrochemical refining, Acid/Caustic handling, biohazard cleanup, tank cleaning, pharmaceutical manufacturing |
Polyethylene, Spunbond Polypropylene | |
Elastic | |
Hooded | |
Bound | |
ASTM F 903, NFPA 99 | |
1.5 mil |