Kimberly-Clark Professional™ V30 Nemesis™ Safety Glasses LC12/2
Kimberly-Clark™ Professional V30 Nemesis™ Safety Glasses LC12/2 have a patented Flex-Dry nose piece designed to be extremely flexible, to channel sweat away from the eyes, and to help prevent slippage down the nose for long-term comfort
Supplier: Kimberly-Clark Professional™ 20379
Description
- Stylish
- Durable
- Sporty, flexible, lightweight design
- Polycarbonate lenses provide 99.9% UVA/ UVB/UVC protection to help prevent eye damage
- Wraparound frame has a base curve of 8.25 that extends the range of peripheral protection
- Soft touch temples for comfort
- Each pair comes with a neck cord
- Manufactured to comply with the ANSI / ISEA Z87.1+ standard for personal eye protection and are CSA certified
- Comfortable and functional for construction, manufacturing, gun shooting / shooting range, and a host of other industries
Specifications
Clear | |
Black | |
12/Cs. |
V30 NEMESIS Protective Spectacle | |
Free neck cord | |
Soft touch |