Kimberly-Clark Professional™ V30 Nemesis™ Safety Glasses LC12/2
Kimberly-Clark™ Professional V30 Nemesis™ Safety Glasses LC12/2 have a patented Flex-Dry nose piece designed to be extremely flexible, to channel sweat away from the eyes, and to help prevent slippage down the nose for long-term comfort
$92.03 - $116.32
Specifications
Product Type | V30 NEMESIS Protective Spectacle |
---|---|
Temple Style | Soft touch |
Catalog Number | Mfr. No. | Frame Color | Lens Tint | Lens Coating | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Frame Color | Lens Tint | Lens Coating | Price | Quantity | |||
19-169-877
|
Kimberly-Clark Professional™
20378 |
Black | Clear | Uncoated |
Case of 12 Each for $92.03
|
|
|||
19-169-878
|
Kimberly-Clark Professional™
20380 |
Black | Smoke | Uncoated |
Case of 12 Each for $106.54
|
|
|||
19-169-880
|
Kimberly-Clark Professional™
20379 |
Black | Clear |
Case of 12 Each for $116.32
|
|
||||
19-169-879
|
Kimberly-Clark Professional™
20381 |
Black | Indoor/Outdoor |
Case of 12 Each for $109.27
|
|
||||
Description
- Stylish
- Durable
- Sporty, flexible, lightweight design
- Polycarbonate lenses provide 99.9% UVA/ UVB/UVC protection to help prevent eye damage
- Wraparound frame has a base curve of 8.25 that extends the range of peripheral protection
- Soft touch temples for comfort
- Each pair comes with a neck cord
- Manufactured to comply with the ANSI / ISEA Z87.1+ standard for personal eye protection and are CSA certified
- Comfortable and functional for construction, manufacturing, gun shooting / shooting range, and a host of other industries
Specifications
V30 NEMESIS Protective Spectacle | |
Soft touch |